Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081906.1 | NP_001075375.1 | 5 - 6 | 749 | 93 | |
GenBank mRNA | AY919871.1 | - | 5 - 6 | 749 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus Inventoried Probe spans exons |