Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081917.1 | NP_001075386.1 | 2 - 3 | 115 | 148 | |
GenBank mRNA | DQ399295.1 | - | 2 - 3 | 115 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Equus caballus Probe spans exons |