Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081920.1 | 4 - 5 | 570 | 70 | ||
XM_014734994.2 | XP_014590480.1 | 673 | 70 | |||
XM_014734995.2 | XP_014590481.1 | 628 | 70 | |||
GenBank mRNA | DQ788676.1 | - | 4 - 5 | 570 | 70 |
Gene Symbol: LOC111769981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626347.1 | XP_023482115.1 | 673 | 70 | ||
XM_023626348.1 | XP_023482116.1 | 632 | 70 |
Gene Symbol: LOC111769983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626350.1 | XP_023482118.1 | 673 | 70 | ||
XM_023626351.1 | XP_023482119.1 | 632 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |