Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081926.2 | NP_001075395.1 | 3 - 4 | 664 | 62 | |
GenBank mRNA | AF126744.1 | - | 3 - 4 | 664 | 62 | |
AF126745.1 | - | 3 - 4 | 664 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |