Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081931.2 | NP_001075400.1 | 1 - 2 | 132 | 66 | |
GenBank mRNA | AJ251189.1 | - | 1 - 2 | 132 | 66 | |
EU438774.1 | - | 1 - 2 | 130 | 66 | ||
EU779497.1 | - | 1 - 2 | 132 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Inventoried Probe spans exons |