Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081937.1 | NP_001075406.1 | 299 | 65 | ||
GenBank mRNA | AY339891.1 | - | 299 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus |