Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082495.2 | NP_001075964.1 | 1 - 2 | 148 | 147 | |
GenBank mRNA | U62529.1 | - | 1 - 2 | 150 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |