Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082504.1 | NP_001075973.1 | 2 - 3 | 448 | 108 | |
GenBank mRNA | X78077.1 | - | 448 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Equus caballus Probe spans exons |