Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILT11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082527.1 | NP_001075996.1 | 5 - 6 | 785 | 72 | |
XM_005596095.2 | XP_005596152.1 | 768 | 72 | |||
GenBank mRNA | AB120409.1 | - | 5 - 6 | 785 | 72 | |
AB120410.1 | - | 4 - 5 | 722 | 72 | ||
AB120411.1 | - | 5 - 6 | 740 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Probe spans exons |