Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099760.1 | NP_001093230.1 | 3 - 4 | 292 | 78 | |
XM_005602598.3 | XP_005602655.2 | 589 | 78 | |||
GenBank mRNA | EF397505.1 | - | 3 - 4 | 292 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |