Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099767.2 | NP_001093237.1 | 2 - 3 | 104 | 117 | |
GenBank mRNA | Y18204.1 | - | 107 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |