Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111299.2 | NP_001104769.2 | 2 - 3 | 958 | 70 | |
GenBank mRNA | EU025848.1 | - | 2 - 3 | 958 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Equus caballus Probe spans exons |