Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130078.1 | NP_001123550.1 | 3 - 4 | 279 | 143 | |
XM_023636871.1 | XP_023492639.1 | 590 | 143 | |||
GenBank mRNA | EU438776.1 | - | 3 - 4 | 279 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Equus caballus Probe spans exons |