Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101652.1 | NP_001095122.1 | 9 | 1650 | 130 | |
GenBank mRNA | EU014893.1 | - | 9 | 1650 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |