Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650222.1 | XP_023505990.1 | 597 | 68 | ||
XM_023650232.1 | XP_023506000.1 | 597 | 68 | |||
XM_023650241.1 | XP_023506009.1 | 472 | 68 | |||
GenBank mRNA | AJ010121.1 | - | 3 - 4 | 407 | 68 | |
JF681042.1 | - | 3 - 4 | 391 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |