Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626834.1 | XP_023482602.1 | 750 | 104 | ||
XM_023626835.1 | XP_023482603.1 | 751 | 104 | |||
GenBank mRNA | AY762637.1 | - | 2 - 3 | 317 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Equus caballus Probe spans exons |