Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142400.1 | NP_001135872.1 | 8 - 9 | 956 | 81 | |
XM_005609945.3 | XP_005610002.1 | 989 | 81 | |||
XM_005609946.3 | XP_005610003.1 | 797 | 81 | |||
XM_023640280.1 | XP_023496048.1 | 978 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |