Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159335.1 | NP_001152807.1 | 1 - 2 | 65 | 85 | |
XM_005606474.3 | XP_005606531.1 | 185 | 85 | |||
XM_014736717.2 | XP_014592203.1 | 135 | 85 | |||
XM_014736718.2 | XP_014592204.1 | 165 | 85 | |||
GenBank mRNA | FM994925.1 | - | 1 - 2 | 65 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |