Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164063.2 | NP_001157535.1 | 2 - 3 | 162 | 86 | |
GenBank mRNA | FN432030.1 | - | 2 - 3 | 164 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus Probe spans exons |