Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163880.1 | NP_001157352.1 | 2 - 3 | 69 | 78 | |
XM_005601758.2 | XP_005601815.1 | 304 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |