Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168402.1 | NP_001161874.1 | 1 - 2 | 230 | 82 | |
GenBank mRNA | CX604538.1 | - | 1 - 2 | 162 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Equus caballus Probe spans exons |