Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081834.2 | NP_001075303.1 | 5 | 923 | 90 | |
NM_001114535.2 | NP_001108007.1 | 5 | 923 | 90 | ||
GenBank mRNA | AB178284.1 | - | 5 | 924 | 90 |
Gene Symbol: LOC111774802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648109.1 | XP_023503877.1 | 1872 | 90 | ||
XM_023648110.1 | XP_023503878.1 | 1841 | 90 | |||
XM_023648111.1 | XP_023503879.1 | 1040 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Equus caballus Both primers and probe map within a single exon |