Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364902.1 | NP_001351831.1 | 7 - 8 | 977 | 100 | |
GenBank mRNA | AJ271386.1 | - | 6 - 7 | 973 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |