Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418869.6 | XP_418869.1 | 1 - 2 | 146 | 94 | |
GenBank mRNA | BX929537.1 | - | 1 - 2 | 103 | 94 | |
BX934915.1 | - | 1 - 2 | 141 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |