Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285926.1 | NP_001272855.1 | 7 - 8 | 608 | 131 | |
XM_004938247.3 | XP_004938304.1 | 7 - 8 | 692 | 131 | ||
XM_004938248.3 | XP_004938305.1 | 7 - 8 | 608 | 131 | ||
GenBank mRNA | BU433840.1 | - | 3 - 4 | 335 | 131 | |
BX930121.1 | - | 7 - 8 | 605 | 131 | ||
BX930142.2 | - | 8 - 9 | 755 | 131 | ||
BX930156.1 | - | 8 - 9 | 755 | 131 | ||
BX930576.2 | - | 7 - 8 | 605 | 131 | ||
BX930906.1 | - | 7 - 8 | 618 | 131 | ||
BX933264.1 | - | 7 - 8 | 588 | 131 | ||
BX935375.1 | - | 7 - 8 | 608 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |