Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151304.1 | XP_025007072.1 | 3 - 4 | 418 | 86 | |
GenBank mRNA | BX930048.1 | - | 2 - 3 | 221 | 86 | |
BX930970.1 | - | 1 - 2 | 327 | 86 | ||
BX931464.1 | - | 2 - 3 | 232 | 86 | ||
BX932516.1 | - | 2 - 3 | 217 | 86 | ||
BX934657.1 | - | 2 - 3 | 240 | 86 | ||
BX935487.2 | - | 2 - 3 | 232 | 86 | ||
BX936079.2 | - | 2 - 3 | 234 | 86 | ||
BX950593.1 | - | 1 - 2 | 314 | 86 | ||
CR385483.1 | - | 2 - 3 | 239 | 86 | ||
CR407467.1 | - | 2 - 3 | 238 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |