Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151304.1 | XP_025007072.1 | 4 - 5 | 531 | 79 | |
GenBank mRNA | BX930048.1 | - | 3 - 4 | 334 | 79 | |
BX930970.1 | - | 2 - 3 | 440 | 79 | ||
BX931464.1 | - | 3 - 4 | 345 | 79 | ||
BX932516.1 | - | 3 - 4 | 330 | 79 | ||
BX934657.1 | - | 3 - 4 | 353 | 79 | ||
BX935487.2 | - | 3 - 4 | 345 | 79 | ||
BX936079.2 | - | 3 - 4 | 347 | 79 | ||
BX950593.1 | - | 2 - 3 | 427 | 79 | ||
CR385483.1 | - | 3 - 4 | 352 | 79 | ||
CR407467.1 | - | 3 - 4 | 351 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |