Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942659.3 | XP_004942716.1 | 4 - 5 | 232 | 69 | |
GenBank mRNA | BX930573.1 | - | 3 - 4 | 219 | 69 | |
BX931475.1 | - | 3 - 4 | 212 | 69 | ||
BX932964.1 | - | 212 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |