Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942659.3 | XP_004942716.1 | 6 - 7 | 477 | 63 | |
GenBank mRNA | BX930573.1 | - | 5 - 6 | 464 | 63 | |
BX931475.1 | - | 5 - 6 | 457 | 63 | ||
BX932964.1 | - | 457 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |