Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942612.3 | XP_004942669.1 | 2 - 3 | 159 | 70 | |
XM_004942613.3 | XP_004942670.1 | 2 - 3 | 159 | 70 | ||
XM_025152676.1 | XP_025008444.1 | 2 - 3 | 159 | 70 | ||
XM_421938.5 | XP_421938.2 | 3 - 4 | 289 | 70 | ||
GenBank mRNA | BX931745.1 | - | 3 - 4 | 227 | 70 | |
BX933154.2 | - | 3 - 4 | 322 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |