Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949575.3 | XP_004949632.1 | 5 - 6 | 606 | 69 | |
XM_015280691.2 | XP_015136177.1 | 5 - 6 | 518 | 69 | ||
XM_424940.6 | XP_424940.5 | 5 - 6 | 593 | 69 | ||
GenBank mRNA | BX932541.1 | - | 5 - 6 | 574 | 69 | |
BX934080.1 | - | 5 - 6 | 593 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |