Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414580.5 | XP_414580.1 | 7 - 8 | 880 | 68 | |
GenBank mRNA | BX934896.1 | - | 4 - 5 | 298 | 68 | |
CR385449.1 | - | 141 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |