Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419050.6 | XP_419050.1 | 7 - 8 | 825 | 77 | |
GenBank mRNA | BX930669.1 | - | 3 - 4 | 355 | 77 | |
BX936222.2 | - | 7 - 8 | 791 | 77 | ||
BX936269.2 | - | 7 - 8 | 791 | 77 | ||
BX950392.1 | - | 7 - 8 | 791 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |