Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277518.1 | NP_001264447.1 | 2 - 3 | 113 | ||
XM_015283466.2 | XP_015138952.1 | 2 - 3 | 113 | |||
XM_015283467.2 | XP_015138953.1 | 2 - 3 | 113 | |||
XM_015283468.2 | XP_015138954.1 | 2 - 3 | 113 | |||
GenBank mRNA | BX929403.2 | - | 2 - 3 | 343 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |