Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277137.1 | NP_001264066.1 | 2 - 3 | 183 | 125 | |
GenBank mRNA | BX933453.2 | - | 2 - 3 | 193 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |