Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198654.1 | NP_001185583.1 | 5 - 6 | 913 | 113 | |
GenBank mRNA | BX929395.2 | - | 5 - 6 | 910 | 113 | |
BX933798.2 | - | 785 | 113 | |||
BX933966.2 | - | 5 - 6 | 913 | 113 | ||
CR407508.1 | - | 3 - 4 | 890 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |