Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277728.1 | NP_001264657.1 | 4 - 5 | 561 | 87 | |
GenBank mRNA | BU105412.1 | - | 3 - 4 | 510 | 87 | |
BU222325.1 | - | 4 - 5 | 563 | 87 | ||
BX935079.1 | - | 3 - 4 | 683 | 87 | ||
BX935284.2 | - | 3 - 4 | 515 | 87 | ||
CR386367.1 | - | 3 - 4 | 606 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |