Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039263.2 | NP_001034352.2 | 5 - 6 | 589 | 86 | |
XM_015294144.2 | XP_015149630.1 | 5 - 6 | 582 | 86 | ||
GenBank mRNA | AJ456913.1 | - | 5 - 6 | 590 | 86 | |
CR387393.1 | - | 5 - 6 | 576 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |