Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015283159.2 | XP_015138645.1 | 16 - 17 | 2379 | 101 | |
XM_025148148.1 | XP_025003916.1 | 15 - 16 | 2751 | 101 | ||
XM_025148149.1 | XP_025003917.1 | 15 - 16 | 2751 | 101 | ||
XM_025148150.1 | XP_025003918.1 | 15 - 16 | 2263 | 101 | ||
XM_418456.6 | XP_418456.2 | 15 - 16 | 2354 | 101 | ||
GenBank mRNA | CR387563.1 | - | 1 - 2 | 146 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |