Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946472.3 | XP_004946529.1 | 3 - 4 | 545 | 68 | |
XM_015295990.2 | XP_015151476.1 | 3 - 4 | 539 | 68 | ||
GenBank mRNA | CR387598.1 | - | 3 - 4 | 322 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |