Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPDPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197037.1 | NP_001183966.1 | 4 - 5 | 395 | 67 | |
GenBank mRNA | BX930759.1 | - | 4 - 5 | 386 | 67 | |
BX931456.2 | - | 4 - 5 | 386 | 67 | ||
BX935797.2 | - | 4 - 5 | 382 | 67 | ||
BX935801.1 | - | 4 - 5 | 390 | 67 | ||
BX935878.2 | - | 4 - 5 | 365 | 67 | ||
BX936190.2 | - | 4 - 5 | 391 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |