Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277881.2 | XP_015133367.1 | 2 | 518 | 134 | |
GenBank mRNA | CR387170.1 | - | 1 | 350 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |