Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199250.1 | NP_001186179.1 | 1 - 2 | 152 | 62 | |
GenBank mRNA | CR390140.1 | - | 1 - 2 | 78 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |