Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FERMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936527.3 | XP_004936584.2 | 17 | 3065 | 172 | |
XM_025150867.1 | XP_025006635.1 | 15 | 2932 | 172 | ||
XM_025150868.1 | XP_025006636.1 | 14 | 2899 | 172 | ||
XM_025150869.1 | XP_025006637.1 | 15 | 3059 | 172 | ||
GenBank mRNA | CR391344.1 | - | 1 | 549 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |