Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STKLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945935.3 | XP_004945992.1 | 13 - 14 | 1861 | 149 | |
XM_004945936.3 | XP_004945993.1 | 13 - 14 | 1858 | 149 | ||
XM_004945937.3 | XP_004945994.1 | 12 - 13 | 1757 | 149 | ||
XM_004945939.3 | XP_004945996.1 | 13 - 14 | 1581 | 149 | ||
XM_025141408.1 | XP_024997176.1 | 13 - 14 | 1575 | 149 | ||
GenBank mRNA | CR523822.1 | - | 2 - 3 | 231 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |