Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305183.1 | NP_001292112.1 | 6 - 7 | 948 | 64 | |
XM_025149776.1 | XP_025005544.1 | 6 - 7 | 904 | 64 | ||
GenBank mRNA | CR353076.1 | - | 6 - 7 | 924 | 64 | |
X54612.1 | - | 693 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |