Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277734.2 | XP_015133220.1 | 1 - 2 | 220 | 103 | |
GenBank mRNA | BX931965.2 | - | 1 - 2 | 137 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |