Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282235.2 | NP_001269164.1 | 3 - 4 | 724 | 86 | |
GenBank mRNA | CR388896.1 | - | 3 - 4 | 725 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |