Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416702.6 | XP_416702.1 | 2 - 3 | 324 | 76 | |
GenBank mRNA | CR353053.1 | - | 2 - 3 | 330 | 76 | |
CR391444.1 | - | 2 - 3 | 333 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |