Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419986.5 | XP_419986.1 | 2 - 3 | 260 | 135 | |
GenBank mRNA | BX931493.1 | - | 2 - 3 | 243 | 135 | |
BX933820.1 | - | 2 - 3 | 233 | 135 | ||
BX936239.1 | - | 2 - 3 | 237 | 135 | ||
BX950504.1 | - | 2 - 3 | 275 | 135 | ||
CR406049.1 | - | 2 - 3 | 241 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |